missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5103667
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Antibody detects endogenous levels of total SLC22A17.
The Slc22 family of organic anion and cation transporters (OATs, OCTs, OCTNs) are transmembrane proteins expressed predominantly in kidney and liver. Each contain 12 predicted alpha-helical transmembrane domains (TMDs) and one large extracellular loop between TMDs 1 and 2. Transporters of the SLC22 family function in different ways such as uniporters that mediate facilitated diffusion in either direction (OCTs), as anion exchangers (OAT1, OAT3 and URAT1), and as Na(+)/l-carnitine cotransporter (OCTN2). Slc22 family members participate in the absorption and/or excretion of drugs, xenobiotics, and endogenous compounds in intestine, liver, and kidney, and perform homeostatic functions in brain and heart. Mutations in the Slc22 family may cause specific diseases such as primary systemic carnitine deficiency or idiopathic renal hypouricemia and may change drug absorption or excretion. Recent studies show the expression of Slc22A17 as receptor for Lipocalin 2 is relatively high in hematopoietic stem cells.
Especificaciones
| SLC22A17 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A17 | |
| 1700094C23Rik; 24p3 receptor; 24p3R; AU041908; AW555662; BOCT; Boit; brain-specific organic ion transporter; brain-type organic cation transporter; hBOIT; Lipocalin-2 receptor; mBOCT; Neutrophil gelatinase-associated lipocalin receptor; NGAL receptor; NgalR; NGALR2; NGALR3; organic cation transporter BOCT; potent brain type organic ion transporter; rBOCT; SLC22A17; solute carrier family 22 (organic cation transporter), member 17; solute carrier family 22 member 17; solute carrier family 22, member 17 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 305886, 51310, 59049 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WUG5, Q9D9E0, Q9P290 | |
| SLC22A17 | |
| A synthesized peptide derived from human SLC22A17(Accession Q8WUG5), corresponding to amino acid residues A483-T533. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido