missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: KLPETLGKRL ATTWEEAAKL ESENESKSSK LLLTTNNSGL EKTEAITPRD SGLGE Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 34%.
This gene encodes a member of the organic zwitterion transporter protein family which transports carnitine. The encoded protein has also been shown to transport anticancer drugs like bleomycin successful treatment has been correlated with the level of activity of this transporter in tumor cells.
Specifications
Specifications
| Antígeno | SLC22A16 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | SLC22A16 |
| N.º de referencia del gen | Q86VW1 |
| Alias de gen | Carnitine transporter 2; CT2; dJ261K5.1; epididymis secretory protein Li 18; flipt 2; FLIPT2; fly-like putative organic ion transporter 2; fly-like putative transporter 2; HEL-S-18; OAT6; OCT6; OKB1; organic cation transporter 6; organic cation transporter OKB1; Organic cation/carnitine transporter 6; SLC22A16; solute carrier family 22 (organic cation transporter), member 16; solute carrier family 22 (organic cation/carnitine transporter), member 16; solute carrier family 22 member 16; WUGSC:RG331P03.1 |
| Símbolos de los genes | SLC22A16 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
