Learn More
SLC1A5/ASCT2 Mouse, anti-Human, Clone: 1C4E1, Proteintech
Código de producto. 18084485 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18084485 20 μL 20 microlitros
18096875 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18084485 Proveedor Proteintech N.º de proveedor 685401IG20UL
Mouse Monoclonal Antibody
The SLC1A5/ASCT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC1A5/ASCT2. This antibody recognizes human antigen. The SLC1A5/ASCT2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
Antígeno SLC1A5/ASCT2
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clon 1C4E1
Concentración 1000 μg/mL
Símbolos de los genes AAAT, ASCT2, ATB(0), ATBO, Baboon M7 virus receptor, FLJ31068, M7V1, M7VS1, R16, RDR, RDRC, SLC1A5
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 6510
Especies diana Human
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG1