Learn More
SLAIN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16832334 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16832334 20 μL 20 microlitros
16822334 150 μL 150 microlitros
2 options
Código de producto. 16832334
Marca: Proteintech 139081AP20UL
Rabbit Polyclonal Antibody
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Especificaciones
Antígeno SLAIN2
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.31 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica SLAIN2
N.º de referencia del gen Q8CI08, Q9P270
Alias de gen KIAA1458, SLAIN motif family, member 2, SLAIN2
Símbolos de los genes SLAIN2
Especie del huésped Rabbit
Inmunógeno SLAIN2 Fusion Protein Ag4888
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 305310, 57606, 75991
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG