Learn More
SIRT1 Rabbit, anti-Human, CoraLite Plus 750, Polyclonal, Proteintech
Código de producto. 18014795 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
20 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
18014795 100 μL 100 microlitros
18030526 20 μL 20 microlitros
2 options
Código de producto. 18014795
Marca: Proteintech CL75013161100UL
Rabbit Polyclonal Antibody
The SIRT1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SIRT1. This antibody recognizes human antigen. The SIRT1 antibody has been validated for the following applications: FC (Intra), WB analysis. This antibody is coralite™ plus 750.
Especificaciones
Antígeno SIRT1
Aplicaciones Western Blot, Flow Cytometry (Intracellular Staining)
Concentración 1000 μg/mL
Conjugado CoraLite Plus 750
Símbolos de los genes hSIR2, hSIRT1, SIR2 like protein 1, SIR2L1, SIRT1
Especie del huésped Rabbit
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 23411
Especies diana Human
Contenido y almacenamiento Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG