Learn More
Thermo Scientific™ Single-Use BioProcess Container, 2D, Aegis™ 5-14, 3 ports
GREENER_CHOICE
Código de producto. 17829786 Tienda Thermo Scientific Productos
Change view
Click to view available options
Capacidad (métrico):
2 L
5 L
10 L
20 L
50 L
Tamaño de la unidad:
paquete de 10
Código de producto. Capacidad (métrico) unitSize
17829786 5 L paquete de 10
17839796 2 L paquete de 10
17839776 10 L paquete de 10
17859786 20 L paquete de 10
17839786 50 L paquete de 10
5 options
Código de producto. 17829786
Marca: Thermo Scientific™ SH31266.01
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
Features:
• All BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Flexible and economic solution to update or increase your bioreactor capacity
• Suitable for both suspension and adherent cultures
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from a large component library – with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and gas filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.
TRUSTED_SUSTAINABILITY
Especificaciones
Capacidad (métrico) 5 L
Cantidad 5 L
Tubing Style TPE