Learn More
Thermo Scientific™ Single-Use BioProcess Container, 2D, Aegis™ 5-14, 2 ports
GREENER_CHOICE
Código de producto. 17829766 Tienda Thermo Scientific Productos
Change view
Click to view available options
Capacidad (métrico):
50 mL
100 mL
250 mL
500 mL
1 L
2 L
3 L
Tamaño de la unidad:
paquete de 10
Código de producto. Capacidad (métrico) unitSize
17829766 250 mL paquete de 10
17899756 50 mL paquete de 10
17809696 100 mL paquete de 10
17839766 500 mL paquete de 10
17899686 1 L paquete de 10
17849776 2 L paquete de 10
17859776 3 L paquete de 10
7 options
Código de producto. 17829766
Marca: Thermo Scientific™ SH31265.13
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
The Thermo Scientific Single-Use BPC is composed of Aegis 5-14 film, a single-web, five-layer, animal-origin free film.
Features:
• All BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
• Manufactured with industry-leading Aegis 5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Flexible and economic solution to update or increase your bioreactor capacity
• Suitable for both suspension and adherent cultures
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from a large component library – with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
• Filters—used for bioburden reduction; sterilization grade filters and gas filters.
• Tubing—thermoplastic elastomers, platinum-cured silicone.
• Sample ports—septum-style and needle-free ports.
• Clamps—sliders, pinch, and crimp bands.
TRUSTED_SUSTAINABILITY
Especificaciones
Capacidad (métrico) 250 mL
Cantidad 250 mL
Tubing Style TPE