missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin1/MAPKAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP1-89569
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sin1/MAPKAP1 Polyclonal specifically detects Sin1/MAPKAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| Sin1/MAPKAP1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| JC310, MEKK2-interacting protein 1, MGC2745, MIP1SIN1SIN1b, Mitogen-activated protein kinase 2-associated protein 1, mitogen-activated protein kinase associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MAPKAP1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:NPFNEYSKFDGKGHVGTTATKKIDVYLPLHSSQDRLLPMTVVTMASARVQDLIGLICWQYTSEGREPKLNDNVSAYCLHIAEDDGEVDTDFPPLDSNEPIHKFGFSTLALVEKYSSPGLTSKESLFVRINAAHGFSLIQVDNTKVTMKE | |
| 0.1 mL | |
| Autophagy, Cancer, mTOR Pathway, Protein Kinase | |
| 79109 | |
| Human, Mouse, Rat | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido