Learn More
Simport™ Scientific Sample Tubes
For transport and storage of urine, sputum and most liquids or particulate samples
Descripción
Descripción
  • Tube made of polypropylene
  • Cap made of polyethylene
  • Conical bottom tubes with self-standing base graduated
  • Chemically resistant and shatterproof
  • Supplied with a leakproof screw cap
  • Molded-in graduations make them easy to read
For transporting liquid sample/hazardous material