Learn More
Fosfato de plata, 99 %, Thermo Scientific Chemicals
Código de producto. p-4407821
Change view
Click to view available options
Cantidad:
5 g
25 g
Tamaño de la unidad:
25 g
5 g
11311239 5 g 5 g
11301239 25 g 25 g
2 options
Código de producto. 11311239
Marca: Thermo Scientific Alfa Aesar 011415.06
Solicitar formato a granel o personalizado
CAS: 7784-09-0 | Ag3O4P | 418.57 g/mol
Silver phosphate finds applications in photography, and in silver staining of biological materials. It is used as a photocatalyst for the visible light photochemical splitting of water, and for production of activated oxygen by the same method. It is employed in Crystal growth for scintillation, optics and dosimetry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 7784-09-0
Fórmula molecular Ag3O4P
Peso molecular (g/mol) 418.57
Número MDL MFCD00014148
Clave InChI FJOLTQXXWSRAIX-UHFFFAOYSA-K
Sinónimo trisilver phosphate, trisilver 1+ phosphate, unii-zl6t4y1xp8, zl6t4y1xp8, silver orthophosphate, trisilver orthophosphate, silver phosphate ag3po4, silberphosphat, trisilver 1+ orthophosphate, silver orthophosphate ag3po4
PubChem CID 159913
Nombre IUPAC triplata;fosfato
SMILES [Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O
Especificaciones
Nombre del producto químico o material Silver phosphate
Punto de fusión 849°C
Color Gris a negro
Forma física Polvo
Cantidad 5 g
Intervalo de porcentaje de ensayo 99%
Información de solubilidad Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3,ammonia,ammonium carbonate,alkali cyanides and thiosulfates.
Formula Weight (peso de la fórmula) 418.58
Olor Odorless
Sensibilidad Light sensitive
Aspecto Yellow powder
Densidad 6.37