missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGLEC7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13939-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Putative adhesion molecule that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,3- and alpha-2,6-linked sialic acid. Also binds disialogangliosides (disialogalactosyl globoside, disialyl lactotetraosylceramide and disialyl GalNAc lactotetraoslylceramide). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. Mediates inhibition of natural killer cells cytotoxicity. May play a role in hemopoiesis. Inhibits differentiation of CD34+ cell precursors towards myelomonocytic cell lineage and proliferation of leukemic myeloid cells (in vitro).Especificaciones
| SIGLEC7 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC7 | |
| AIRM 1, AIRM1, CD328, CDw328, D siglec, p75, p75/AIRM1, QA79, QA79 membrane protein, SIGLEC 7, SIGLEC7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27036 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y286 | |
| SIGLEC7 | |
| SIGLEC7 Fusion Protein Ag4220 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido