missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-F Rat anti-Mouse , Alexa Fluor 488, Clone: E50-2440, BD Biosciences™
Rat Monoclonal Antibody
Marca: BD Biosciences 567005
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Siglec-F Monoclonal antibody specifically detects Siglec-F in Mouse samples. It is validated for Flow Cytometry
Especificaciones
| Siglec-F | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Siglecf; Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5 | |
| Mouse Siglec-F and human IgG Fc recombinant fusion protein | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| E50-2440 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 488 under optimum conditions, and unreacted Alexa Fluor 488 was removed. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido