missing translation for 'onlineSavingsMsg'
Learn More

Siglec-10 Antibody (5G6) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30160668
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30160668 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30160668 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP321494JF669

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Siglec-10 Monoclonal antibody specifically detects Siglec-10 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Siglec-10
Aplicaciones ELISA, Immunohistochemistry, Immunoprecipitation
Clasificación Monoclonal
Clon 5G6
Conjugado Janelia Fluor 669
Formulación 50mM Sodium Borate
Alias de gen MGC126774, PRO940, sialic acid binding Ig-like lectin 10, sialic acid-binding Ig-like lectin 10, SIGLEC-10, siglec-like gene 2, Siglec-like protein 2, SLG2sialic acid binding Ig-like lectin 10 Ig-like lectin 7
Especie del huésped Mouse
Inmunógeno Siglec-10 extracellular domain fused with human IgG1 Fc fragment
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 89790
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.