Learn More
SHOWA S-TEX KV3 Cut-Resistant Knit Gloves
Click to view available options
Tamaño:
Pequeño
M
L
XL
Tamaño de la unidad:
10 pares
Descripción
- The glove has a natural rubber palm dip that ensures a superior grip in both dry and wet conditions.
- For maximum cut resistance, the KV3 glove is made with a combination of stainless steel liner, Kevlar fibers, and patented Hagane Coil technology.
- The KV3 glove is designed to be form-fitting and breathable, ensuring maximum comfort and performance during use.
Especificaciones
Especificaciones
| Estructura | Seamless Knit |
| Coating Material | Látex |
| Color | Negro, Yellow |
| Hand | Específico |
| Latex Free | No |
| Material | Aramid Knit, Polyester, Stainless Steel Fiber |
| Tipo de producto | Cut-resistant Gloves |
| Tamaño | Pequeño |
| Sexo | Unisex |
| Estilo de puño de los guantes | Knit |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction