Learn More
SHOWA 4561 Nitrile Knit Gloves
Descripción
- The sponge nitrile palm coating provides a strong grip, even in wet or greasy environments.
- The seamless knit reduces irritation, and its elasticated, form-fitting design keeps dirt out.
Especificaciones
Especificaciones
| Estructura | Seamless Knit |
| Coating Material | Nitrilo |
| Color | Yellow, Negro |
| Hand | Específico |
| Latex Free | Sí |
| Material | Nitrilo, Kevlar |
| Tipo de producto | Cut-resistant Gloves |
| Tamaño | M |
| Sexo | Unisex |
| Estilo de puño de los guantes | Knit |