Learn More
SH2B3 Rabbit anti-Human, Polyclonal, Proteintech
Código de producto. 16811735 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16811735 20 μL 20 microlitros
16801735 150 μL 150 microlitros
2 options
Código de producto. 16811735
Marca: Proteintech 221981AP20UL
Rabbit Polyclonal Antibody
This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.
Especificaciones
Antígeno SH2B3
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica SH2B3
N.º de referencia del gen Q9UQQ2
Alias de gen LNK, Lymphocyte adapter protein, SH2B adapter protein 3, SH2B adaptor protein 3, SH2B3
Símbolos de los genes SH2B3
Especie del huésped Rabbit
Inmunógeno SH2B3 Fusion Protein Ag17381
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10019
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG