missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA533020
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SGK3 Polyclonal Antibody for IHC (P)
SGKL (SGK3) is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Especificaciones
| SGK3 | |
| Polyclonal | |
| Unconjugated | |
| SGK3 | |
| 2510015P22Rik; A330005P07Rik; CISK; cytokine-independent survival kinase; DKFZp781N0293; fy; fz; Serine/threonine-protein kinase Sgk3; serum glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase family member 3; serum/glucocorticoid regulated kinase family, member 3; serum/glucocorticoid-regulated kinase 3; Serum/glucocorticoid-regulated kinase-like; SGK2; SGK3; Sgkl | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23678 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q96BR1 | |
| SGK3 | |
| Synthetic 17 amino acid peptide from C-terminus of human SGK3. | |
| 50 μg | |
| Primary | |
| Human, Rabbit, Monkey | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido