missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFTPD Polyclonal Antibody

Descripción
Immunogen sequence: RDGRDGREGP RGEKGDPGLP GAAGQAGMPG QAGPVGPKGD NGSVGEPGPK GDTGPSGPPG PPGVPGPAGR EGPLGKQGNI GPQGKPGPKG EAGPKGEVGA PGMQGSAGAR GLAGPKGERG VPGERGVPGN TGAAGSAGAM GPQGSPGARG PPGLKGDKGI PGDKGAKGES G.
Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.
Especificaciones
Especificaciones
| Antígeno | SFTPD |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.48 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| génica | SFTPD |
| N.º de referencia del gen | P35247 |
| Alias de gen | AI573415; COLEC7; Collectin7; collectin-7; CP4; EGK_19692; Lung surfactant protein D; PSPD; PSP-D; PSP-D Surfactant protein D; pulmonary surfactant apoprotein; Pulmonary surfactant associated protein PSP-D; pulmonary surfactant protein D; pulmonary surfactant-associated protein D; SFTP 4; Sftp4; SFTPD; SPD; SP-D; surfactant associated protein D; Surfactant associated protein pulmonary 4; surfactant protein D; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4 |
| Símbolos de los genes | SFTPD |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?