missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SFRS8 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15716631 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15716631 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15716631 Proveedor Invitrogen™ N.º de proveedor PA558754

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: NYLHPSLFAS KKCNRLEELM KPLKVVDPDH PLAALVRKAQ ADSSTPTPHN ADGAPVQPSQ VEYTADSTVA AMYYSYYMLP DGTYCLAPPP PGIDVTTY Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SFRS8
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica SFSWAP
N.º de referencia del gen Q12872
Alias de gen 1190005N23Rik; 6330437E22Rik; AI197402; AW212079; serine/arginine-rich splicing factor 8; Sfrs8; SFSWAP; splicing factor SWAP homolog; splicing factor, arginine/serine-rich 8; splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, Drosophila); splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, Drosophila homolog); splicing factor, suppressor of white-apricot family; splicing factor, suppressor of white-apricot homolog; splicing factor, suppressor of white-apricot homolog (Drosophila); Srsf8; Suppressor of white apricot protein homolog; SWAP
Símbolos de los genes SFSWAP
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human SFRS8. Recombinant protein control fragment (Product #RP-97660).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6433
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.