Learn More
SFPQ Mouse anti-Human, Unconjugated, Clone: 1G4A5, Proteintech
Código de producto. 16889575 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16889575 20 μL 20 microlitros
16879575 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16889575 Proveedor Proteintech N.º de proveedor 671291IG20UL
Mouse Monoclonal Antibody
The SFPQ antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SFPQ. This antibody recognizes human, mouse, rat antigen. The SFPQ antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Antígeno SFPQ
Aplicaciones Western Blot, ELISA
Clasificación Monoclonal
Clon 1G4A5
Concentración 500 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 6421
Especies diana Human, Rat, Mouse
Isotype IgG1