missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: MSTVMSRKGP APELQGVEVA LAPEELELDP MAMTQKYEEH VREQQAQVEK EDFSDMVAEH AAKQKQKKRK AQPQDSRGGS KKYKEFK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Subunit of the splicing factor SF3B required for 'A' complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA. May also be involved in the assembly of the 'E' complex. Belongs also to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron.
Specifications
Specifications
| Antígeno | SF3B2 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.05 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | SF3B2 |
| N.º de referencia del gen | Q13435 |
| Alias de gen | 145kDa; 2610311M13Rik; 2810441F20Rik; B230398H18Rik; Cus1; pre-mRNA splicing factor SF3b 145 kDa subunit; pre-mRNA-splicing factor SF3b 145 kDa subunit; SAP 145; SAP145; SF3b1; SF3B145; SF3b150; SF3B2; spliceosome associated protein 145; spliceosome-associated protein 145; Splicing factor 3B subunit 2; splicing factor 3b, subunit 2; splicing factor 3b, subunit 2, 145kD; splicing factor 3b, subunit 2, 145kDa |
| Símbolos de los genes | SF3B2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
