Learn More
Invitrogen™ SETD8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA580762
Descripción
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SETD8/PR-Set7.
SETD8 is a histone methyltransferase that specifically monomethylates 'Lys-20' of histone H4. H4 'Lys-20' monomethylation is enriched during mitosis and represents a specific tag for epigenetic transcriptional repression. The protein mainly functions in euchromatin regions, thereby playing a central role in the silencing of euchromatic genes. It is required for cell proliferation, probably by contributing to the maintenance of proper higher order structure of DNA during mitosis. It is also involved in chromosome condensation and proper cytokinesis. Nucleosomes are preferred as substrate compared to free histones.
Especificaciones
| SETD8 | |
| Polyclonal | |
| PBS with no preservative | |
| Q9NQR1 | |
| KMT5A | |
| Recombinant Human SETD8/PR-Set7 Protein (Lys195-His352). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| KMT5A | |
| 2410195B05Rik; AA617402; AW536475; H4-K20-HMTase; H4-K20-HMTase KMT5A; H4-K20-HMTase SETD8; H4-K20-specific histone methyltransferase; H4K20-specific histone methyltransferase splice variant Set8b; histone methyltransferase Pr-set7/Set8; histone-lysine N-methyltransferase KMT5A; histone-lysine N-methyltransferase SETD8; KMT5A; lysine (K)-specific methyltransferase 5A; lysine methyltransferase 5A; lysine N-methyltransferase 5A; Lysine-specific methylase 5A; N-lysine methyltransferase KMT5A; N-lysine methyltransferase SETD8; PR/SET domain containing protein 8; PR/SET domain-containing protein 07; PR/SET07; PRSET7; PR-Set7; RGD1305893; SET domain containing (lysine methyltransferase) 8; SET domain containing protein 8 variant 1; SET domain containing protein 8 variant 2; SET domain-containing 8; SET domain-containing protein 8; SET07; SET8; SETD8; Unknown (protein for MGC:134426) | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 387893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.