missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Especificaciones
| Antígeno | SETD2 |
|---|---|
| Concentración | 0.58 mg/mL |
| Aplicaciones | Western Blot, Immunofluorescence, Immunocytochemistry |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
16896835
|
Proteintech
55377-1-AP-20UL |
20 μL |
145.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
16886835
|
Proteintech
55377-1-AP-150UL |
150 μL |
410.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
This antibody is specific to SETD2.
Histone methyltransferase that specifically trimethylates ′Lys-36′ of histone H3 (H3K36me3) using dimethylated ′Lys-36′ (H3K36me2) as substrate. Represents the main enzyme generating H3K36me3, a specific tag for epigenetic transcriptional activation. Plays a role in chromatin structure modulation during elongation by coordinating recruitment of the FACT complex and by interacting with hyperphosphorylated POLR2A. Acts as a key regulator of DNA mismatch repair in G1 and early S phase by generating H3K36me3, a mark required to recruit MSH6 subunit of the MutS alpha complex: early recruitment of the MutS alpha complex to chromatin to be replicated allows a quick identification of mismatch DNA to initiate the mismatch repair reaction. H3K36me3 also plays an essential role in the maintece of a heterochromatic state, by recruiting DNA methyltransferase DNMT3A. H3K36me3 is also enhanced in intron-containing genes, suggesting that SETD2 recruitment is enhanced by splicing and that splicing is coupled to recruitment of elongating RNA polymerase. Required during angiogenesis. Recruited to the promoters of adenovirus 12 E1A gene in case of infection, possibly leading to regulate its expression.Especificaciones
| SETD2 | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9BYW2 | |
| 29072 | |
| Peptide | |
| Primary | |
| -20°C | |
| SETD2 |
| 0.58 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HIF 1, HIF1, HIP 1, hSET2, HSPC069, Huntingtin yeast partner B, HYPB, KIAA1732, KMT3A, Lysine N methyltransferase 3A, p231HBP, SET domain containing 2, SET2, SETD2 | |
| SETD2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto