Learn More
SETD2 Monoclonal Antibody (OTI3G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca:  Origene Technologies TA804268
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17583773
  • 540.87€ / 100 microlitros
Descripción
Histone methyltransferase that specifically trimethylates ′Lys-36′ of histone H3 (H3K36me3) using dimethylated ′Lys-36′ (H3K36me2) as substrate. Represents the main enzyme generating H3K36me3, a specific tag for epigenetic transcriptional activation. Plays a role in chromatin structure modulation during elongation by coordinating recruitment of the FACT complex and by interacting with hyperphosphorylated POLR2A. Acts as a key regulator of DNA mismatch repair in G1 and early S phase by generating H3K36me3, a mark required to recruit MSH6 subunit of the MutS alpha complex: early recruitment of the MutS alpha complex to chromatin to be replicated allows a quick identification of mismatch DNA to initiate the mismatch repair reaction. H3K36me3 also plays an essential role in the maintece of a heterochromatic state, by recruiting DNA methyltransferase DNMT3A. H3K36me3 is also enhanced in intron-containing genes, suggesting that SETD2 recruitment is enhanced by splicing and that splicing is coupled to recruitment of elongating RNA polymerase. Required during angiogenesis. Recruited to the promoters of adenovirus 12 E1A gene in case of infection, possibly leading to regulate its expression.
Especificaciones
Especificaciones
SETD2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BYW2
SETD2
Human recombit protein fragment corresponding to amino acids 1787-2144 of human SETD2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI3G8
Unconjugated
SETD2
HBP231, HIF-1, HIP-1, HSPC069, HYPB, KMT3A, LLS, SET2, p231HBP
Mouse
Affinity Chromatography
RUO
29072
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones