missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERTAD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-81542
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SERTAD2 Polyclonal specifically detects SERTAD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| SERTAD2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| KIAA0127, MGC126688, Sei-2, SERTA domain containing 2, SERTA domain-containing protein 2, Transcriptional regulator interacting with the PHD-bromodomain 2, transcriptional regulator interacting with the PHS-bromodomain 2, TRIP-Br2MGC126690 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SERTAD2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DGPSKVSYTLQRQTIFNISLMKLYNHRPLTEPSLQKTVLINNMLRRIQEELKQEGSLRPMFTPSSQPTTEPSDSYREAPPAFSHLASPSSHPCDLGSTTPLEACLTPASLLEDDDDTFCTSQ | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 9792 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido