missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINF2 Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA547101
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
In Western blots, approximately 15% cross-reactivity with recombinant human (rh) Serpin F2 and less than 1% cross-reactivity with recombinant mouse (rm) Serpin C1, rmSerpin D1, rhSerpin A1, rmSerpin A5, and rhSerpin C1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
| SERPINF2 | |
| Polyclonal | |
| Unconjugated | |
| SERPINF2 | |
| A2AP; AAP; AI747498; alpha 2 antiplasmin; alpha-2-antiplasmin; Alpha-2-AP; alpha-2-PI; Alpha-2-plasmin inhibitor; API; pigment epithelium derived factor; plasmin inhibitor alpha 2; Pli; serine (or cysteine) peptidase inhibitor, clade F, member 2; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; serine (or cysteine) proteinase inhibitor, clade F, member 2; Serpimf2; serpin F2; serpin family F member 2; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; Serpinf2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 18816 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q61247 | |
| SERPINF2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Serpin F2/alpha 2-Antiplasmin Val28-Lys491. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido