missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SerpinB9 Antibody (7D8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB100-64359B
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SerpinB9 Monoclonal antibody specifically detects SerpinB9 in Human, Mouse (Negative), Porcine (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Especificaciones
| SerpinB9 | |
| Monoclonal | |
| Biotin | |
| CAP-3, CAP3PI-9, Cytoplasmic antiproteinase 3, Peptidase inhibitor 9, PI9protease inhibitor 9 (ovalbumin type), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9, serpin B9, serpin peptidase inhibitor, clade B (ovalbumin), member 9, serpin peptidase inhibitor, clade B, member 9 | |
| Recombinant PI-9 produced in P. pastoris. | |
| 0.1 mL | |
| Apoptosis, Immunology | |
| 5272 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 7D8 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Porcine (Negative) | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido