missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINB8 Polyclonal Antibody

Descripción
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse SERPINB8/SerpinB8.
The superfamily of high molecular weight serine proteinase inhibitors. A critical portion of the molecule, the reactive center loop connects beta sheets A and C. Protease inhibitor-8 (PI8; SERPINB8) is a member of the ov-serpin subfamily, which, relative to the archetypal serpin PI1 (MIM 107400), is characterized by a high degree of homology to chicken ovalbumin, lack of N- and C-terminal extensions, absence of a signal peptide, and a serine rather than an asparagine residue at the penultimate position.
Especificaciones
Especificaciones
| Antígeno | SERPINB8 |
| Aplicaciones | ELISA, Immunoprecipitation, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | SERPINB8 |
| N.º de referencia del gen | O08800 |
| Alias de gen | CAP2; CAP-2; Cytoplasmic antiproteinase 2; NK10; ovalbumin; peptidase inhibitor 8; PI8; PI-8; protease inhibitor 8 (ovalbumin type); serine (or cysteine) peptdiase inhibitor, clade B, member 8; serine (or cysteine) peptidase inhibitor, clade B, member 8; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 8; serine (or cysteine) proteinase inhibitor, clade B, member 8; serine protease inhibitor 8; serpin B8; serpin family B member 8; serpin peptidase inhibitor, clade B (ovalbumin), member 8; Serpinb8; Spi8 |
| Símbolos de los genes | SERPINB8 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?