Descripción
This gene encodes an alpha-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
Especificaciones
Especificaciones
| Antígeno | SERPINA6 |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.25 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | SERPINA6 |
| N.º de referencia del gen | P08185 |
| Alias de gen | CBG, Serpin A6, SERPINA6, Transcortin |
| Símbolos de los genes | SERPINA6 |
| Mostrar más |