missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINA1 Mouse anti-Canine, Human, Clone: OTI5B12, lyophilized, TrueMAB™
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin, which may be released during inflammatory reactions in the lung. In the absence of alpha-1-AT, these enzymes are not inhibited and they may digest pulmonary parenchyma. Alpha-1-AT deficiency is associated with chronic obstructive lung disease (emphysema) and less frequently with hepatic cirrhosis in infants and respiratory distress of the newborn. Increase in alpha-1-AT occurs as an acute phase response to tissue necrosis and inflammation. Serum level of alpha-1-AT is elevated in rheumatoid arthritis, bacterial infections, vasculitis, and carcinomatosis.
Especificaciones
Especificaciones
| Antígeno | SERPINA1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI5B12 |
| Conjugado | Unconjugated |
| Formulación | PBS with 8% trehalose and no preservative; pH 7.3 |
| génica | SERPINA1 |
| N.º de referencia del gen | P01009 |
| Alias de gen | A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT, nNIF |
| Símbolos de los genes | SERPINA1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?