missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin F1/PEDF Antibody (OTI1F7), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-74119V
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Human, Mouse samples. It is validated for Western Blot.
Especificaciones
| Serpin F1/PEDF | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SERPINF1 | |
| Full length recombinant protein of human Serpin F1/PEDF (NP_002606) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2a |
| Western Blot | |
| OTI1F7 | |
| Western Blot | |
| Cell proliferation-inducing gene 35 protein, EPC-1PIG35, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Neuroscience, Vision | |
| 5176.0 | |
| Store at 4°C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido