missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERCA2 ATPase Recombinant Rabbit Monoclonal Antibody (10F0M9)

Descripción
SERCA2 ATPase Recombinant Monoclonal Antibody for Western Blot, ICC/IF, ELISA, IP
This gene encodes one of the SERCA Ca(2+)-ATPases, which are intracellular pumps located in the sarcoplasmic or endoplasmic reticula of muscle cells. This enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol into the sarcoplasmic reticulum lumen, and is involved in regulation of the contraction/relaxation cycle. Mutations in this gene cause Darier-White disease, also known as keratosis follicularis, an autosomal dominant skin disorder characterized by loss of adhesion between epidermal cells and abnormal keratinization. Alternative splicing results in multiple transcript variants encoding different isoforms.
Especificaciones
Especificaciones
| Antígeno | SERCA2 ATPase |
| Aplicaciones | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 10F0M9 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | Atp2a2 |
| N.º de referencia del gen | O55143, P11507, P16615 |
| Alias de gen | 9530097L16Rik; ATP2; Atp2a2; ATP2B; ATPase Ca++ transporting cardiac muscle slow twitch 2; ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2; ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2; ATPase, Ca++ transporting, cardiac muscle, slow twitch 2; ATPase, Ca++ transporting, slow twitch 2; Ca(2+)-transport ATPase class 3; calcium ATPase; calcium pump 2; calcium-ATPase (EC 3.6.1.3); calcium-transporting ATPase; calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; cardiac Ca2+ ATPase; D5Wsu150e; DAR; DD; endoplasmic reticulum class 1/2 Ca(2+) ATPase; mKIAA4195; putative SERCA isoform; sarco(endo)plasmic reticulum Ca(2+)-dependent ATPase 2; sarco/endoplasmic reticulum Ca2+-ATPase 2; sarcoplasmic reticulum Ca2+-transport ATPase isoform; sarcoplasmic/endoplasmic reticulum calcium ATPase 2; sarcoplasmic/endoplasmic-reticulum Ca(2+) pump gene 2; SERCA; SERCA ATPase; SERCA2; Serca2a; SERCA2B; SercaII; SR Ca(2+)-ATPase 2; unnamed protein product |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?