missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Septin-7 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15772681 Tienda Thermo Scientific Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
15772681 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15772681

Marca: Invitrogen™ PA554755

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: MSVSARSAAA EERSVNSSTM VAQQKNLEGY VGFANLPNQV YRKSVKRGFE FTLMVVGESG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Septin-7
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
génica SEPTIN7
N.º de referencia del gen O55131, Q16181, Q9WVC0
Alias de gen 7-Sep; Cdc10; CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 protein homolog; CDC3; cell division cycle 10 homolog; E430034N22; NBLA02942; SEPT7; SEPT7A; septin 7; septin 7 variant 4; Septin7; septin-7
Símbolos de los genes SEPTIN7
Especie del huésped Rabbit
Inmunógeno Recombinant protein corresponding to Human Septin-7. Recombinant protein control fragment (Product #RP-92690).
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 235072, 64551, 989
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.