missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody is specific to SENP5.
SUMO (small ubiquitin-related modifier), a member of the ubiquitin-like protein family, regulates diverse cellular functions of a variety of target proteins including transcription, DNA repair, nucleocytoplasmic trafficking and chromosome segregation. SUMO precursor proteins undergo cleavage of the residues after the "GG" region by SUMO-specific proteases in maturation. This cleavage of the precursor is a prerequisite for subsequent sumoylation. The sentrin-specific (or SUMO-specific) protease (SENP) proteins belong to the peptidase C48 family and include SENP1-3 and SENP5-8. SENP1, SENP2 and SENP3 degrade UBL1 and SMT3H2 conjugates and subsequently release the monomers from sumoylated substrates.
Specifications
Specifications
| Antígeno | SENP5 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.49 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | SENP5 |
| N.º de referencia del gen | Q6NXL6, Q96HI0 |
| Alias de gen | SENP5, Sentrin specific protease 5 |
| Símbolos de los genes | Senp5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?