Learn More
SELK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16800334 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16800334 150 μL 150 microlitros
16810334 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16800334 Proveedor Proteintech N.º de proveedor 125611AP150UL
Rabbit Polyclonal Antibody
SELK is a selenoprotein, containing a selenocysteine residue at its active site. SELK is localized to the endoplasmic reticulum and is highly expressed in the heart, where it may function as an antioxidant. SELK can bind to proteins such as the ER-associated degradation (ERAD) components p97 ATPase, Derlin-1 and -2, and SelS as well as proteins in the oligosaccharyltransferase complex, suggesting that SELK may be involved in the Derlin-dependent ERAD of glycosylated misfolded proteins. SELK is also thought to be important in Ca2+ flux in immune cells, and is cleaved by m-calpain in resting macrophages. When these macrophages are activated through different Toll-like receptors, this cleavage is inhibited
Especificaciones
Antígeno SELK
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.15 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica SELENOK
N.º de referencia del gen P59798, Q9JLJ1, Q9Y6D0
Alias de gen HSPC030, HSPC297, selenoprotein K, SELK
Símbolos de los genes Selenok
Especie del huésped Rabbit
Inmunógeno SELK Fusion Protein Ag3143
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 290549, 58515, 80795
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG