Learn More
Secretogranin II Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Tienda Proteintech ProductosDescripción
The Secretogranin II antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Secretogranin II. This antibody recognizes human, mouse, rat antigen. The Secretogranin II antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | Secretogranin II |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA |
| Clasificación | Polyclonal |
| Concentración | 233 μg/mL |
| Conjugado | Unconjugated |
| Especie del huésped | Rabbit |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 7857 |
| Especies diana | Human, Rat, Mouse |
| Mostrar más |