missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC22B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14776-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.Especificaciones
| SEC22B | |
| Polyclonal | |
| Unconjugated | |
| SEC22B | |
| ER Golgi SNARE of 24 kDa, ERS 24, ERS24, SEC22B, SEC22L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20333, 310710, 9554 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08547, O75396, Q4KM74 | |
| Sec22b | |
| SEC22B Fusion Protein Ag6077 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido