Learn More
SDR42E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17347-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| SDR42E1 | |
| Polyclonal | |
| Unconjugated | |
| SDR42E1 | |
| HSPC105, SDR42E1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 307897, 74032, 93517 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WUS8, Q9D665 | |
| Sdr42e1 | |
| SDR42E1 Fusion Protein Ag11391 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |