missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SDHC Polyclonal Antibody

Descripción
SDHC Polyclonal Antibody for Western Blot
This gene encodes one of four nuclear-encoded subunits that comprise succinate dehydrogenase, also known as mitochondrial complex II, a key enzyme complex of the tricarboxylic acid cycle and aerobic respiratory chains of mitochondria. The encoded protein is one of two integral membrane proteins that anchor other subunits of the complex, which form the catalytic core, to the inner mitochondrial membrane. There are several related pseudogenes for this gene on different chromosomes. Mutations in this gene have been associated with paragangliomas. Alternatively spliced transcript variants have been described.
Especificaciones
Especificaciones
| Antígeno | SDHC |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.7 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.09% sodium azide; pH 7.4 |
| génica | SDHC |
| N.º de referencia del gen | Q99643, Q9CZB0 |
| Alias de gen | 0610010E03Rik; AI316496; AU019277; CYB560; CYBL; cytochrome B large subunit of complex II; integral membrane protein CII-3; integral membrane protein CII-3b; large subunit of cytochrome b; PGL3; QPs1; QPs-1; SDH3; SDHC; succinate dehydrgenase cytochrome b; succinate dehydrogenase 3, integral membrane subunit; succinate dehydrogenase complex subunit C; succinate dehydrogenase complex subunit C integral membrane protein 15kDa; succinate dehydrogenase complex, subunit C, integral membrane protein; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kD; succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa; succinate dehydrogenase cytochrome b560 subunit, mitochondrial; Succinate-ubiquinone oxidoreductase cytochrome B large subunit |
| Símbolos de los genes | SDHC |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?