Descripción
Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis.
Especificaciones
Especificaciones
| Antígeno | SDHB |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.13 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | SDHB |
| N.º de referencia del gen | P21912, P21913, Q9CQA3 |
| Alias de gen | IP, PGL4, SDH, SDH1, SDHB, SDHIP |
| Símbolos de los genes | SDHB |
| Mostrar más |