Learn More
SDHB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16827113 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16827113 150 μL 150 microlitros
16837113 20 μL 20 microlitros
2 options
Código de producto. 16827113
Marca: Proteintech 106201AP150UL
Rabbit Polyclonal Antibody
Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis.
Especificaciones
Antígeno SDHB
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica SDHB
N.º de referencia del gen P21912, P21913, Q9CQA3
Alias de gen IP, PGL4, SDH, SDH1, SDHB, SDHIP
Símbolos de los genes SDHB
Especie del huésped Rabbit
Inmunógeno SDHB Fusion Protein Ag0968
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 298596, 6390, 67680
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG