missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SDHA Recombinant Rabbit Monoclonal Antibody (JM10-83)
GREENER_CHOICE

Código de producto. 16386004
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16386004 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16386004 Proveedor Invitrogen™ N.º de proveedor MA532598

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes a major catalytic subunit of succinate-ubiquinone oxidoreductase, a complex of the mitochondrial respiratory chain. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. Mutations in this gene have been associated with a form of mitochondrial respiratory chain deficiency known as Leigh Syndrome. A pseudogene has been identified on chromosome 3q29.
TRUSTED_SUSTAINABILITY

Tekniske data

Antígeno SDHA
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Clasificación Recombinant Monoclonal
Clon JM10-83
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
génica SDHA
N.º de referencia del gen P31040, Q7ZVF3, Q8K2B3, Q920L2
Alias de gen 1500032O14Rik; 2310034D06Rik; 4921513A11; C81073; CMD1GG; Flavoprotein subunit of complex II; FP; im:7141001; mitochondrial succinate dehydrogenase complex subunit A; PGL5; SDH1; SDH2; SDHA; SDHA protein; SDHF; SDHFP1; si:dkeyp-84f11.2; succinate dehydrogenase [ubiquinone] flavoprotein subunit; succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex flavoprotein subunit A; succinate dehydrogenase complex subunit A; succinate dehydrogenase complex subunit A flavoprotein; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp); succinate dehydrogenase flavoprotein subunit; succinate dehydrogenase flavoprotein subunit A; zgc:56051
Símbolos de los genes SDHA
Especie del huésped Rabbit
Inmunógeno Synthetic peptide within Human SDHA aa 581-624.
Método de purificación Protein A
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 157074, 393884, 6389, 66945
Especies diana Human, Mouse, Rat, Zebrafish
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.