Descripción
Silencer for Labconco Scroll Pumps filters exhaust while silencing exhaust pump noise. It also prevents tip seal dust from contaminating the environment. Sound absorbent element is replaceable. Includes filter, centering ring and clamp to attach to exhaust outlet.
Especificaciones
Especificaciones
| Peso con embalaje (imperial) | 2.0 lb. |
| Peso con embalaje (métrico) | 0.9 kg |
| Incluye | Filter, Centering ring and Clamp to attach to exhaust outlet |
| Para utilizar con | Bomba de vacío con paletas giratorias |
| Tipo de bomba | Vacuum Pump |