missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN9A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567651
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Predicted to react with Human samples.
SCN9A Polyclonal antibody specifically detects SCN9A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Especificaciones
| SCN9A | |
| Polyclonal | |
| Unconjugated | |
| SCN9A | |
| ETHA; FEB3B; GEFSP7; hNaN; hNE Na; hNE-Na; HSAN2D; KIAA1356; Kiaa4197; MED; mKIAA4197; NAC1; NAC2; NAC3; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.6; Nav1.7; Nav1.8; Nav1.9; NE; NE NA; NENA; NE-NA; neuroendocrine sodium channel; Peripheral sodium channel 1; PN1; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; Scn2a; SCN2A1; SCN2A2; SCN3A; SCN4A; SCN5A; SCN8A; Scn9a; SFNP; SNS2; sodium channel 25; sodium channel protein type 9 subunit alpha; Sodium channel protein type IX subunit alpha; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage-gated, type 9, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; sodium voltage-gated channel alpha subunit 9; voltage-gated sodium channel alpha subunit Nav1.7; voltage-gated sodium channel subunit alpha Nav1.7 | |
| Rabbit | |
| affinity chromatography | |
| RUO | |
| 6335 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15858 | |
| SCN9A | |
| Synthesized peptide derived from human SCN9A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido