missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCN5A Polyclonal Antibody
GREENER_CHOICE

Código de producto. 17158273
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
17158273 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17158273

Marca: Invitrogen™ PA5110346

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Immunogen sequence: YVMSENFSRP LGPPSSSSIS STSFPPSYDS VTRATSDNLQ VRGSDYSHSE DLADFPPSPD RDRESIV.

Scn5a is an integral membrane protein and tetrodotoxin-resistant voltage-gated sodium channel subunit. Scn5a is found primarily in cardiac muscle and is responsible for the initial upstroke of the action potential in an electrocardiogram. Defects in Scn5a are a cause of long QT syndrome type 3 (LQT3), an autosomal dominant cardiac disease. Alternative splicing results in several transcript variants encoding different isoforms of Scn5a. Voltage-gated sodium channesl (VGSCs) mediate regenerative cell membrane depolarization and conduction of electrical signalling in nerves and muscles. Expression of Scn5a is also detected in lymphocytes, glia, and fibroblasts. Further, Scn5a mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, Scn5a forms a sodium-selective channel through which Na(+) ions may pass in accordance with their electrochemical gradient. Functionally, Scn5a is responsible for the initial upstroke of the action potential and the channel inactivation is regulated by intracellular calcium levels. Voltage-gated sodium channels (NaV) are responsible for action potential initiation and propagation in excitable cells, including nerve, muscle, and neuroendocrine cell types. Scn5a are also expressed at low levels in non-excitable cells, where their physiological role is unclear. Diseases associated with SCN5A include Long Qt Syndrome-3 and Brugada Syndrome 1.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SCN5A
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 1.74 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
génica SCN5A
N.º de referencia del gen Q14524, Q9JJV9
Alias de gen cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CDCD2; CDCD2VF1; CIN5; CMD1E; CMPD2; HB1; HB2; HBBD; HH1; HH1CMD1E; ICCD; IVF; LQT3; LQT3SSS1CMPD2; mH1; Nav1.5; Nav1.5c; PFHB1; RATRSKM2X; RH1; RSkM2; RSKM2X; SCAL; Scn2x; SCN5A; SkM1; SkM2; sodium channel protein cardiac muscle subunit alpha; Sodium channel protein type 5 subunit alpha; sodium channel protein type V subunit alpha; sodium channel voltage-gated type V alpha polypeptide; sodium channel, voltage gated, type V alpha subunit; sodium channel, voltage-gated, type 5, alpha subunit; sodium channel, voltage-gated, type V, alpha; sodium channel, voltage-gated, type V, alpha polypeptide; sodium channel, voltage-gated, type V, alpha subunit; sodium voltage-gated channel alpha subunit 5; SSS1; VF1; voltage-gated sodium channel Nav1.5c; voltage-gated sodium channel subunit alpha Nav1.5
Símbolos de los genes SCN5A
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to a sequence within amino acids 1950-2016 of human SCN5A (NP_932173.1).
Método de purificación Affinity Chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 20271, 6331
Especies diana Human, Mouse
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.