missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCF/c-kit Ligand Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-27162SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Human samples. It is validated for Western Blot.
Especificaciones
| SCF/c-kit Ligand | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| KITLG | |
| A synthetic peptide corresponding to amino acids 121-158 of mouse kitl was used as immunogen, GenBank no gb AAA39378.1 . | |
| 0.025 mg | |
| Biologically Active Proteins, Cancer, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 4254 | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 3-5ug/ml | |
| DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in mouse and 77% homologous in rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido