Learn More
Sca1 (Ly-6A/E) Rat, anti-Mouse, PE, Clone: E13-161.7, STEMCELL Technologies
Rat Monoclonal Antibody
Marca: Stemcell Technologies 60032PE
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- The E13-161.7 antibody reacts with Sca1 (Stem Cell Antigen-1 or Ly-6A/E), an 18 kDa GPI-linked protein belonging to the lymphocyte activation protein-6 (Ly-6) family
- Sca1 is expressed on the surface of hematopoietic stem and progenitor cells, myeloid cells, and peripheral B and T lymphocytes
- Sca1 expression levels are strongly upregulated in all strains upon cellular activation
- Sca1 is involved in the regulation of T and B cell responses and is believed to play roles in the differentiation, proliferation, and survival of a variety of stem cells
- Sca1 has emerged as a phenotypic marker of choice for identifying and isolating hematopoietic stem and progenitor cells
Especificaciones
| Sca1 (Ly-6A/E) | |
| Monoclonal | |
| PE | |
| 200 μg | |
| Mouse | |
| Hematopoietic Stem and Progenitor Cells, Mammary Cells |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| E13-161.7 | |
| Rat | |
| Primary | |
| IgG2a κ |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.