missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SC4MOL Polyclonal Antibody
Descripción
Peptide sequence: MATNESVSIF SSASLAVEYV DSLLPENPLQ EPFKNAWNYM LNNYTKFQIA Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 79%.
Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases.Sterol-C4-mehtyl oxidase-like protein was isolated based on its similarity to the yeast ERG25 protein. It contains a set of putative metal binding motifs with similarity to that seen in a family of membrane desaturases-hydroxylases. The protein is localized to the endoplasmic reticulum membrane and is believed to function in cholesterol biosynthesis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | SC4MOL |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | MSMO1 |
| N.º de referencia del gen | Q15800 |
| Alias de gen | 1500001G16Rik; C-4 methylsterol oxidase; C-4 methylsterol oxidase-like protein; C78600; DESP4; ERG25; MCCPD; Methylsterol monooxygenase 1; methylsterol monoxygenase 1; MGC104344; MSMO1; Neuropep 1; neurorep 1; RANP-1; sc4mol; sterol-C4-methyl oxidase; sterol-C4-methyl oxidase-like; sterol-C4-methyl oxidase-like protein; wu:fb59g06; wu:fb66e06; zgc:56437 |
| Símbolos de los genes | MSMO1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?