missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB10540-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Spike RBD Monoclonal specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Western Blot.
Especificaciones
| SARS-CoV-2 Spike RBD | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein | |
| Recombinant SARS-CoV-2 Spike Protein, S1 Subunit, Receptor Binding Domain (RBD), Arg319-Phe541, Accession # YP_009724390 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 1034522 | |
| Western Blot 2 ug/mL | |
| YP_009724390 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 43740568 | |
| SARS-CoV-2 | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido