missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Spike Protein S1 Chimeric Recombinant Human Monoclonal Antibody (H6)
Descripción
Recombinant anti-SARS-CoV-2 spike Mouse ScFv is expressed from 293 cells(HEK293) with a human IgG1 Fc tag on C-terminal. Compatible for use in Colloidal Gold Immunochromatography.
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The Spike protein is the main surface antigen of the coronavirus. The glycosylated Spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
Especificaciones
Especificaciones
| Antígeno | SARS-CoV-2 Spike Protein S1 Chimeric |
| Aplicaciones | ELISA, Lateral Flow, Neutralization, Surface Plasmon Resonance |
| Clasificación | Recombinant Monoclonal |
| Clon | H6 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Alias de gen | Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 |
| Especie del huésped | Human |
| Inmunógeno | Recombinant Human Novel Coronavirus Spike glycoprotein (S) (16-685aa). |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?